Assay Details
Gene Symbol: DDX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066212.1 | XP_017921701.1 | 4 - 5 | 1492 | 60 | |
XM_018066213.1 | XP_017921702.1 | 4 - 5 | 1489 | 60 | ||
GenBank mRNA | JO426731.1 | - | 4 - 5 | 60 | ||
KA306779.1 | - | 4 - 5 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
