Assay Details
| Gene Symbol: SESN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053093.1 | XP_017908582.1 | 2 - 3 | 1269 | 60 | |
| XM_018053094.1 | XP_017908583.1 | 2 - 3 | 537 | 60 | ||
| GenBank mRNA | JO598547.2 | - | 8 - 9 | 1962 | 60 | |
More Information
Set Membership: |
|

SESN1
Genomic Map