Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040868.1 | XP_017896357.1 | 4 - 5 | 3144 | 68 | |
| XM_018040876.1 | XP_017896365.1 | 4 - 5 | 2795 | 68 | ||
| XM_018040883.1 | XP_017896372.1 | 3 - 4 | 2663 | 68 | ||
| XM_018040891.1 | XP_017896380.1 | 3 - 4 | 2635 | 68 | ||
| XM_018040901.1 | XP_017896390.1 | 3 - 4 | 2588 | 68 | ||
| GenBank mRNA | JT656522.1 | - | 6 - 7 | 2893 | 68 | |
More Information
Set Membership: |
|

SH3BP4
Genomic Map