Assay Details
Gene Symbol: VIRMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689233.3 | XP_005689290.1 | 21 - 22 | 4883 | 64 | |
XM_005689234.3 | XP_005689291.1 | 20 - 21 | 4730 | 64 | ||
XM_018058232.1 | XP_017913721.1 | 17 - 18 | 4351 | 64 | ||
GenBank mRNA | JT707671.1 | - | 21 - 22 | 4903 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
