Assay Details
| Gene Symbol: LYRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697575.3 | XP_005697632.2 | 3 - 4 | 356 | 80 | |
| XM_005697576.3 | XP_005697633.2 | 4 - 5 | 540 | 80 | ||
| XM_005697577.3 | XP_005697634.2 | 4 - 5 | 527 | 80 | ||
| XM_005697578.3 | XP_005697635.2 | 2 - 3 | 325 | 80 | ||
| XM_005697579.3 | XP_005697636.2 | 4 - 5 | 478 | 80 | ||
| XM_005697580.3 | XP_005697637.2 | 3 - 4 | 496 | 80 | ||
| XM_005697581.3 | XP_005697638.2 | 3 - 4 | 369 | 80 | ||
| XM_018040857.1 | XP_017896346.1 | 2 - 3 | 326 | 80 | ||
| GenBank mRNA | JO620017.2 | - | 2 - 3 | 189 | 80 | |
More Information
Set Membership: |
|

LYRM1
Genomic Map