Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042421.1 | XP_017897910.1 | 8 - 9 | 1353 | 54 | |
| XM_018042432.1 | XP_017897921.1 | 8 - 9 | 1356 | 54 | ||
| XM_018042436.1 | XP_017897925.1 | 7 - 8 | 1187 | 54 | ||
| GenBank mRNA | JO645251.2 | - | 8 - 9 | 1393 | 54 | |
More Information
Set Membership: |
|

RIMS3
Genomic Map