Assay Details
| Gene Symbol: HS3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974991.2 | XP_013830445.2 | 1 - 2 | 904 | 94 | |
| GenBank mRNA | JO617548.2 | - | 1 - 2 | 593 | 94 | |
More Information
Set Membership: |
|

HS3ST2
Genomic Map