Assay Details
| Gene Symbol: LHX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055722.1 | XP_017911211.1 | 2 - 3 | 299 | 90 | |
| XM_018055723.1 | XP_017911212.1 | 2 - 3 | 299 | 90 | ||
| XM_018055724.1 | XP_017911213.1 | 2 - 3 | 128 | 90 | ||
More Information
Set Membership: |
|

LHX6
Genomic Map