Assay Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060945.1 | XP_017916434.1 | 2 - 3 | 2360 | 69 | |
| XM_018060946.1 | XP_017916435.1 | 2 - 3 | 2359 | 69 | ||
| XM_018060947.1 | XP_017916436.1 | 2 - 3 | 2360 | 69 | ||
More Information
Set Membership: |
|

PATZ1
Genomic Map