Assay Details
| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700620.3 | XP_005700677.3 | 2 - 3 | 761 | 67 | |
| GenBank mRNA | JO425258.1 | - | 2 - 3 | 672 | 67 | |
More Information
Set Membership: |
|

SLC16A2
Genomic Map