Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052064.1 | XP_017907553.1 | 3 - 4 | 1241 | 101 | |
| XM_018052066.1 | XP_017907555.1 | 2 - 3 | 1089 | 101 | ||
| XM_018052067.1 | XP_017907556.1 | 3 - 4 | 1242 | 101 | ||
| XM_018052068.1 | XP_017907557.1 | 3 - 4 | 634 | 101 | ||
More Information
Set Membership: |
|

ABHD17B
Genomic Map