Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285603.1 | NP_001272532.1 | 1 | 2316 | 61 | |
| XM_013970466.2 | XP_013825920.2 | 2 | 2553 | 61 | ||
| XM_018060889.1 | XP_017916378.1 | 2 | 2544 | 61 | ||
| GenBank mRNA | DQ872435.1 | - | 1 | 2316 | 61 | |
| GU984768.1 | - | 1 | 2316 | 61 | ||
| JR138558.1 | - | 5 | 745 | 61 | ||
| JT709644.1 | - | 7 | 933 | 61 | ||
More Information
Set Membership: |
|

TLR2
Genomic Map