Assay Details
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285729.1 | NP_001272658.1 | 98 | 96 | ||
| GenBank mRNA | GQ148781.1 | - | 98 | 96 | ||
| JO419477.2 | - | 1945 | 96 | |||
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043685.1 | XP_017899174.1 | 1 - 2 | 222 | 96 | |
More Information
Set Membership: |
|

OTC
Genomic Map