Assay Details
Gene Symbol: PRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285735.1 | NP_001272664.1 | 3 - 4 | 375 | 105 | |
GenBank mRNA | AB231296.1 | - | 3 - 4 | 375 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
