Assay Details
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314182.1 | NP_001301111.1 | 5 - 6 | 356 | 96 | |
XM_018058074.1 | XP_017913563.1 | 6 - 7 | 480 | 96 | ||
GenBank mRNA | JO429519.2 | - | 5 - 6 | 647 | 96 | |
JQ417403.1 | - | 5 - 6 | 356 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
