Assay Details
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285715.1 | NP_001272644.1 | 1 | 619 | 60 | |
| XM_018039349.1 | XP_017894838.1 | 3 | 810 | 60 | ||
| GenBank mRNA | JO609915.2 | - | 3 | 799 | 60 | |
| JQ308184.1 | - | 1 | 619 | 60 | ||
| JT686655.1 | - | 3 | 810 | 60 | ||
| KM881705.1 | - | 3 | 780 | 60 | ||
More Information
Set Membership: |
|

NGF
Genomic Map