Assay Details
| Gene Symbol: TCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285752.1 | NP_001272681.1 | 1 - 2 | 131 | 62 | |
| GenBank mRNA | JN129836.1 | - | 1 - 2 | 131 | 62 | |
More Information
Set Membership: |
|

TCAP
Genomic Map