Assay Details
| Gene Symbol: HSPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314303.1 | NP_001301232.1 | 1 | 1620 | 79 | |
| GenBank mRNA | JN656106.1 | - | 1 | 1620 | 79 | |
| JT707119.1 | - | 1 | 1798 | 79 | ||
| JT710388.1 | - | 1 | 1815 | 79 | ||
More Information
Set Membership: |
|

HSPA2
Genomic Map