Assay Details
| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285636.1 | NP_001272565.1 | 4 - 5 | 371 | 70 | |
| GenBank mRNA | EU847283.1 | - | 4 - 5 | 371 | 70 | |
| EU847288.1 | - | 4 - 5 | 371 | 70 | ||
| HQ326239.1 | - | 4 - 5 | 356 | 70 | ||
| KJ817181.1 | - | 4 - 5 | 371 | 70 | ||
| KP310921.1 | - | 4 - 5 | 371 | 70 | ||
More Information
Set Membership: |
|

FSHR
Genomic Map