Assay Details
| Gene Symbol: CSN1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285695.1 | NP_001272624.1 | 5 - 6 | 168 | 84 | |
| XM_018049127.1 | XP_017904616.1 | 6 - 7 | 287 | 84 | ||
| XM_018049128.1 | XP_017904617.1 | 6 - 7 | 287 | 84 | ||
| XM_018049129.1 | XP_017904618.1 | 6 - 7 | 287 | 84 | ||
| XM_018049130.1 | XP_017904619.1 | 6 - 7 | 287 | 84 | ||
| XM_018049131.1 | XP_017904620.1 | 5 - 6 | 254 | 84 | ||
| XM_018049132.1 | XP_017904621.1 | 6 - 7 | 287 | 84 | ||
| XM_018049133.1 | XP_017904622.1 | 6 - 7 | 287 | 84 | ||
| GenBank mRNA | AY344966.1 | - | 5 - 6 | 168 | 84 | |
| AY530757.1 | - | 4 - 5 | 104 | 84 | ||
| JX047868.1 | - | 5 - 6 | 168 | 84 | ||
| KA319597.1 | - | 6 - 7 | 219 | 84 | ||
| KT253649.1 | - | 5 - 6 | 165 | 84 | ||
| KT253650.1 | - | 5 - 6 | 165 | 84 | ||
| KT253651.1 | - | 5 - 6 | 165 | 84 | ||
| KT253652.1 | - | 5 - 6 | 165 | 84 | ||
| KT253653.1 | - | 5 - 6 | 165 | 84 | ||
| KT253654.1 | - | 5 - 6 | 165 | 84 | ||
| KT253655.1 | - | 5 - 6 | 165 | 84 | ||
| X59836.1 | - | 6 - 7 | 218 | 84 | ||
| X72221.1 | - | 195 | 84 | |||
More Information
Set Membership: |
|

CSN1S1
Genomic Map