Assay Details
| Gene Symbol: IGFBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285566.1 | NP_001272495.1 | 2 - 3 | 561 | 103 | |
| XM_018054265.1 | XP_017909754.1 | 2 - 3 | 1371 | 103 | ||
| GenBank mRNA | JF720883.1 | - | 2 - 3 | 561 | 103 | |
| JO538184.2 | - | 2 - 3 | 1141 | 103 | ||
More Information
Set Membership: |
|

IGFBP5
Genomic Map