Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691832.3 | XP_005691889.2 | 3 - 4 | 857 | 59 | |
| GenBank mRNA | JO413399.2 | - | 2 - 3 | 583 | 59 | |
| KA337161.1 | - | 2 - 3 | 417 | 59 | ||
More Information
Set Membership: |
|

HSD17B2
Genomic Map