Assay Details
| Gene Symbol: ASF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053018.1 | XP_017908507.1 | 3 - 4 | 819 | 74 | |
| GenBank mRNA | JO609319.2 | - | 1 - 2 | 377 | 74 | |
More Information
Set Membership: |
|

ASF1A
Genomic Map