Assay Details
| Gene Symbol: AHSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675161.3 | XP_005675218.1 | 5 - 6 | 822 | 68 | |
| GenBank mRNA | JO420107.2 | - | 1 - 2 | 104 | 68 | |
More Information
Set Membership: |
|

AHSG
Genomic Map