Assay Details
| Gene Symbol: TMEM61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678361.3 | XP_005678418.1 | 2 - 3 | 827 | 84 | |
| XM_018044596.1 | XP_017900085.1 | 3 - 4 | 896 | 84 | ||
More Information
Set Membership: |
|

TMEM61
Genomic Map