Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063401.1 | XP_017918890.1 | 1 - 2 | 467 | 80 | |
| XM_018063402.1 | XP_017918891.1 | 1 - 2 | 493 | 80 | ||
More Information
Set Membership: |
|

HCST
Genomic Map