Assay Details
| Gene Symbol: MRPL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686871.3 | XP_005686928.1 | 2 - 3 | 138 | 61 | |
| GenBank mRNA | KA338500.1 | - | 2 - 3 | 388 | 61 | |
More Information
Set Membership: |
|

MRPL33
Genomic Map