Assay Details
| Gene Symbol: GNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695934.3 | XP_005695991.3 | 5 - 6 | 826 | 55 | |
| XM_013973538.2 | XP_013828992.2 | 5 - 6 | 826 | 55 | ||
| XM_018067129.1 | XP_017922618.1 | 5 - 6 | 826 | 55 | ||
More Information
Set Membership: |
|

GNAT1
Genomic Map