Assay Details
| Gene Symbol: KLK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971438.2 | XP_013826892.2 | 5 - 6 | 662 | 65 | |
| XM_018062881.1 | XP_017918370.1 | 5 - 6 | 720 | 65 | ||
| GenBank mRNA | JT746377.1 | - | 1 - 2 | 347 | 65 | |
| KA350368.1 | - | 4 - 5 | 687 | 65 | ||
More Information
Set Membership: |
|

KLK14
Genomic Map