Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679531.3 | XP_005679588.1 | 3 - 4 | 390 | 110 | |
| GenBank mRNA | JO512646.2 | - | 3 - 4 | 389 | 110 | |
More Information
Set Membership: |
|

MRPS33
Genomic Map