Assay Details
| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683384.3 | XP_005683441.2 | 4 - 5 | 768 | 75 | |
| GenBank mRNA | JO595426.2 | - | 4 - 5 | 736 | 75 | |
More Information
Set Membership: |
|

ZCCHC9
Genomic Map