Assay Details
Gene Symbol: RILPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691362.3 | XP_005691419.2 | 2 - 3 | 867 | 59 | |
XM_005691363.3 | XP_005691420.1 | 2 - 3 | 867 | 59 | ||
XM_018061364.1 | XP_017916853.1 | 2 - 3 | 867 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
