Assay Details
| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695648.2 | XP_005695705.1 | 2 - 3 | 259 | 63 | |
| GenBank mRNA | JO369355.2 | - | 2 - 3 | 266 | 63 | |
More Information
Set Membership: |
|

LMCD1
Genomic Map