Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700174.3 | XP_005700231.2 | 3 - 4 | 949 | 72 | |
| GenBank mRNA | JO420272.2 | - | 2 - 3 | 744 | 72 | |
| JO420275.2 | - | 2 - 3 | 744 | 72 | ||
More Information
Set Membership: |
|

SERPINA7
Genomic Map