Assay Details
| Gene Symbol: TMEM88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972202.2 | XP_013827656.1 | 1 - 2 | 247 | 71 | |
| GenBank mRNA | KA313336.1 | - | 1 - 2 | 292 | 71 | |
More Information
Set Membership: |
|

TMEM88
Genomic Map