Assay Details
| Gene Symbol: NGRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694982.3 | XP_005695039.2 | 2 - 3 | 922 | 107 | |
| GenBank mRNA | JO632469.2 | - | 1 - 2 | 1060 | 107 | |
More Information
Set Membership: |
|

NGRN
Genomic Map