Assay Details
| Gene Symbol: PHF21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048892.1 | XP_017904381.1 | 6 - 7 | 1226 | 65 | |
| XM_018048893.1 | XP_017904382.1 | 6 - 7 | 909 | 65 | ||
More Information
Set Membership: |
|

PHF21B
Genomic Map