Assay Details
| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062939.1 | XP_017918428.1 | 4 - 5 | 1123 | 79 | |
| GenBank mRNA | JO350416.2 | - | 3 - 4 | 552 | 79 | |
More Information
Set Membership: |
|

HAS1
Genomic Map