Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680265.3 | XP_005680322.1 | 1 - 2 | 337 | 60 | |
| GenBank mRNA | JT624901.1 | - | 7 - 8 | 1209 | 60 | |
More Information
Set Membership: |
|

TSPAN31
Genomic Map