Assay Details
| Gene Symbol: MFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680678.3 | XP_005680735.3 | 7 - 8 | 1194 | 72 | |
| GenBank mRNA | JO612314.2 | - | 7 - 8 | 1098 | 72 | |
More Information
Set Membership: |
|

MFNG
Genomic Map