Assay Details
| Gene Symbol: HIF1AN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698330.3 | XP_005698387.1 | 4 - 5 | 762 | 69 | |
| GenBank mRNA | JO318798.2 | - | 4 - 5 | 2343 | 69 | |
| JO397257.2 | - | 4 - 5 | 793 | 69 | ||
More Information
Set Membership: |
|

HIF1AN
Genomic Map