Assay Details
| Gene Symbol: ARHGAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066215.1 | XP_017921704.1 | 3 - 4 | 4278 | 93 | |
| XM_018066216.1 | XP_017921705.1 | 3 - 4 | 4136 | 93 | ||
| XM_018066217.1 | XP_017921706.1 | 3 - 4 | 4102 | 93 | ||
| XM_018066218.1 | XP_017921707.1 | 3 - 4 | 4275 | 93 | ||
| GenBank mRNA | JO559218.2 | - | 4 - 5 | 533 | 93 | |
More Information
Set Membership: |
|

ARHGAP5
Genomic Map