Assay Details
| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675113.3 | XP_005675170.1 | 3 - 4 | 418 | 81 | |
| XM_013963779.2 | XP_013819233.1 | 3 - 4 | 419 | 81 | ||
| XM_018046595.1 | XP_017902084.1 | 3 - 4 | 419 | 81 | ||
| GenBank mRNA | JO449880.2 | - | 2 - 3 | 177 | 81 | |
More Information
Set Membership: |
|

PLAAT1
Genomic Map