Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050141.1 | XP_017905630.1 | 5 - 6 | 639 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF300| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050141.1 | XP_017905630.1 | 5 - 6 | 639 | 102 | |
Set Membership: |
|