Assay Details
| Gene Symbol: NDUFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043457.1 | XP_017898946.1 | 1 - 2 | 345 | 115 | |
| GenBank mRNA | JO393743.2 | - | 2 - 3 | 332 | 115 | |
| JO412706.2 | - | 2 - 3 | 333 | 115 | ||
More Information
Set Membership: |
|

NDUFC2
Genomic Map