Assay Details
| Gene Symbol: MRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061659.1 | XP_017917148.1 | 4 - 5 | 564 | 73 | |
| XM_018061660.1 | XP_017917149.1 | 4 - 5 | 564 | 73 | ||
| GenBank mRNA | JI861637.1 | - | 4 - 5 | 408 | 73 | |
| KA318812.1 | - | 7 - 8 | 2458 | 73 | ||
More Information
Set Membership: |
|

MRS2
Genomic Map