Assay Details
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675642.3 | XP_005675699.2 | 1 - 2 | 144 | 63 | |
XM_018051739.1 | XP_017907228.1 | 1 - 2 | 166 | 63 | ||
GenBank mRNA | JO599303.2 | - | 1 - 2 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
