Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686079.3 | XP_005686136.1 | 1 - 2 | 82 | 62 | |
| XM_013967305.2 | XP_013822759.1 | 1 - 2 | 82 | 62 | ||
More Information
Set Membership: |
|

ISCA2
Genomic Map