Assay Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688804.3 | XP_005688861.1 | 2 - 3 | 214 | 77 | |
| GenBank mRNA | JO364403.2 | - | 1 - 2 | 511 | 77 | |
| KA306106.1 | - | 1 - 2 | 581 | 77 | ||
More Information
Set Membership: |
|

LY6D
Genomic Map