Assay Details
| Gene Symbol: TMEM104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063859.1 | XP_017919348.1 | 9 - 10 | 917 | 58 | |
| XM_018063860.1 | XP_017919349.1 | 9 - 10 | 914 | 58 | ||
| GenBank mRNA | JO426170.1 | - | 1 - 2 | 2467 | 58 | |
More Information
Set Membership: |
|

TMEM104
Genomic Map